Anti-6xHis Mouse Monoclonal Antibody (DyLight® 800) [clone: 33D10.D2]

Lieferant: Rockland Immunochemicals
200-345-382
ROCK200-345-382EA 791 EUR
ROCK200-345-382
Anti-6xHis Mouse Monoclonal Antibody (DyLight® 800) [clone: 33D10.D2]
Antikörper
Type: Primary
Antigen: 6xHis
Clonality: Monoclonal
Clone: 33D10.D2
Conjugation: DyLight® 800
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR