Anti-DYKDDDDK Mouse Monoclonal Antibody (DyLight® 800) [clone: 29E4.G7]
Lieferant: Rockland Immunochemicals
200-345-383
ROCK200-345-383EA 742 EUR
ROCK200-345-383
Anti-DYKDDDDK Mouse Monoclonal Antibody (DyLight® 800) [clone: 29E4.G7]
Antikörper
Type: Primary
Antigen: DYKDDDDK
Clonality: Monoclonal
Clone: 29E4.G7
Conjugation: DyLight® 800
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Bestellen