Anti-GFP Mouse Monoclonal Antibody (DyLight® 405) [clone: 9F9.F9]
Lieferant: Rockland Immunochemicals
200-346-215
ROCK200-346-215EA 791 EUR
ROCK200-346-215
Anti-GFP Mouse Monoclonal Antibody (DyLight® 405) [clone: 9F9.F9]
Antikörper
The emission spectra for this DyLight® conjugate match the principle output wavelengths of most common fluorescence instrumentation.
Recommended Dilutions: ELISA: 1:20000-1:40000; FLISA: >1:20000; Immunofluorescence: >1:5000; Western Blot: 1:10000-1:25000;
Restore with deionized water (or equivalent); 100 µl
Type: Tag
Antigen: GFP
Clonality: Monoclonal
Clone: 9F9.F9
Conjugation: DyLight® 405
Epitope: Full length
Host: Mouse
Isotype: IgG1 kappa
Reactivity: pan