Anti-CPB1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-401-049S
ROCK200-401-049SEA 226 EUR
ROCK200-401-049S
Anti-CPB1 Rabbit Polyclonal Antibody
Antikörper
Anti-Carboxypeptidase B antibody is suitable for ELISA, Western Blot and Immunohistochemistry applications. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500- 1:2000; Immunochemistry: 1:250 - 1:1000; Other Dilution: User Optimized

Type: Primary
Antigen: CPB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig, Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR