Anti-CAT Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-401-051
ROCK200-401-051EA 821 EUR
ROCK200-401-051
Anti-CAT Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:20000; Western Blot: 1:10000 - 1:50000;

Type: Primary
Antigen: CAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine, Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR