Anti-Streptavidin Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-401-095S
ROCK200-401-095SEA 226 EUR
ROCK200-401-095S
Anti-Streptavidin Rabbit Polyclonal Antibody
Antikörper
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:200,000; Western Blot: 1:3000 - 1:12000; Immunochemistry: 1:1500 - 1:5000;

Type: Tag
Antigen: Streptavidin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Streptomyces avidinii
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR