Anti-RFX5 Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
200-401-191
ROCK200-401-191EA 821 EUR
ROCK200-401-191
Anti-RFX5 Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-RFX5 (C-terminal) Reacts with Human, Mouse, Rat
Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;
Type: Primary
Antigen: RFX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human