Anti-UBB Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-401-431S
ROCK200-401-431SEA 226 EUR
ROCK200-401-431S
Anti-UBB Rabbit Polyclonal Antibody
Antikörper
This purified polyclonal antibody reacts with ubiquitin by ELISA and western blot. Although not tested, this antibody is likely functional in immunochemistry and immunoprecipitation. For detection of free Ub by western blotting use Tris-Tricine SDS-PAGE.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:200 - 1:1000;

Type: Primary
Antigen: UBB
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR