Anti-SGK1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-401-844
ROCK200-401-844EA 774 EUR
ROCK200-401-844
Anti-SGK1 Rabbit Polyclonal Antibody
Antikörper
SKG-1 Antibody antibody has been tested for use in ELISA, immunochemistry and western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band at ~49 kDa in size corresponding to SGK-1 by western blotting.

Recommended Dilutions: ELISA: 1:4000 - 1:16000; Western Blot: 1:500 - 1:3000; Immunochemistry: 10 µg/ml to 20 µg/ml

Type: Primary
Antigen: SGK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR