Anti-CFLAR Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-401-983
ROCK200-401-983EA 944 EUR
ROCK200-401-983
Anti-CFLAR Rabbit Polyclonal Antibody
Antikörper
FLIP Antibody has been tested for use in ELISA, western blotting, Immunofluorescence, FACS, and immunochemistry.  HeLa whole cell lysate can be used as a positive control in western blotting, and a band at approximately 55 kDa is expected.

Recommended Dilutions: Western Blot: 0.5-1 µg/ml; Immunochemistry: 10 µg/ml;

Type: Primary
Antigen: CFLAR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR