Anti-SEPHS2 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-401-999
ROCK200-401-999EA 821 EUR
ROCK200-401-999
Anti-SEPHS2 Rabbit Polyclonal Antibody
Antikörper
This Protein A purified antibody has been tested for use in ELISA and western blotting.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 48 kDa in size corresponding to SPS2 by western blotting.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100;

Type: Primary
Antigen: SEPHS2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR