Anti-SARS-CoV Nucleocapsid Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-401-A50
ROCK200-401-A50EA 906 EUR
ROCK200-401-A50
Anti-SARS-CoV Nucleocapsid Rabbit Polyclonal Antibody
Antikörper
This Protein A purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 46 kDa in size corresponding to SARS Nucleocapsid (N) Protein.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:2000 - 1:10000
Restore with deionized water (or equivalent); 500 µl

Type: Primary
Antigen: SARS-CoV Nucleocapsid
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: SARS
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR