Anti-MAP1LC3A Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-401-H57
ROCK200-401-H57EA 913 EUR
ROCK200-401-H57
Anti-MAP1LC3A Rabbit Polyclonal Antibody
Antikörper
Anti-ATG8 antibody is suitable for use in IHC (p) and WB. Expect a band approximately 16 kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 0.25-1 µg/ml; Immunochemistry: 5-10 µg/ml

Type: Primary
Antigen: MAP1LC3A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR