Anti-SMAD6 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-401-I15
ROCK200-401-I15EA 957 EUR
ROCK200-401-I15
Anti-SMAD6 Rabbit Polyclonal Antibody
Antikörper
Anti-SMAD6 antibody is suitable for use in IHC and WB. Expect a band approximately 53kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 1-3 µg/ml; Immunochemistry: 5-10 µg/ml

Type: Primary
Antigen: SMAD6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR