Anti-SMAD7 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-401-I16
ROCK200-401-I16EA 913 EUR
ROCK200-401-I16
Anti-SMAD7 Rabbit Polyclonal Antibody
Antikörper
Anti-SMAD7 antibody is suitbale for use in WB. Expect a band approximately 46kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Western Blot: 2-5 µg/ml

Type: Primary
Antigen: SMAD7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR