Anti-WNVM Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-401-I37
ROCK200-401-I37EA 913 EUR
ROCK200-401-I37
Anti-WNVM Rabbit Polyclonal Antibody
Antikörper
Anti-West Nile (WN) glycoprotein M Antibody is suitbale for use in IF/ICC, IHC (p), IP and WB. Expect a band approximately 8kDa on specific lysates. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: Immunofluorescence: 1-2 µg/ml; Western Blot: 0.5-2 µg/ml; Immunochemistry: 1.0 µg/ml

Type: Primary
Antigen: WNVM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR