Anti-ADH Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-4143-0100
ROCK200-4143-0100EA 821 EUR
ROCK200-4143-0100
Anti-ADH Rabbit Polyclonal Antibody
Antikörper
Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:2000; Immunoprecipitation: 1:100

Type: Primary
Antigen: ADH
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Yeast
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR