Anti-DLD Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-4160-0100
ROCK200-4160-0100EA 821 EUR
ROCK200-4160-0100
Anti-DLD Rabbit Polyclonal Antibody
Antikörper
Anti-LIPOAMIDE DEHYDROGENASE has been assayed against 1.0 ug of Lipoamide Dehydrogenase [Porcine Heart] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG [H&L] (Goat) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100

Type: Primary
Antigen: DLD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR