Anti-L-asparaginase II Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-4171S
ROCK200-4171SEA 226 EUR
ROCK200-4171S
Anti-L-asparaginase II Rabbit Polyclonal Antibody
Antikörper
Anti-L-Asparaginase antibody has been assayed against 1.0 ug of Asparaginase [E. coli] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG [H&L] (Goat) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000; Immunoprecipitation: 1:100

Type: Primary
Antigen: L-asparaginase II
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: E. coli
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR