Anti-MPO Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-4181
ROCK200-4181EA 906 EUR
ROCK200-4181
Anti-MPO Rabbit Polyclonal Antibody
Antikörper
This antibody is suitable for western blotting, ELISA and immunoprecipitation.

Recommended Dilutions: ELISA: 1:5000 - 1:20000; Western Blot: 1:500 - 1:5000; Immunoprecipitation: 1:100;
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Primary
Antigen: MPO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR