Anti-KTI3 Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
200-4187S
ROCK200-4187SEA 226 EUR
ROCK200-4187S
Anti-KTI3 Rabbit Polyclonal Antibody
Antikörper
TRYPSIN INHIBITOR antibody has been assayed against 1.0 ug of Trypsin Inhibitor [Lima Bean] in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG [H&L] (Goat) and ABTS (2,2’-azino-bis-[3-ethylbenthiazoline-6-sulfonic acid])
Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000
Type: Primary
Antigen: KTI3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Lima Bean