Anti-StreptAvidin Rabbit Polyclonal Antibody (Texas Red®)
Lieferant: Rockland Immunochemicals
200-4995
ROCK200-4995EA 5860 EUR
ROCK200-4995
Anti-StreptAvidin Rabbit Polyclonal Antibody (Texas Red®)
Antikörper
Type: Tag
Antigen: StreptAvidin
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pan