Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 53-6.7]
Lieferant: Rockland Immunochemicals
200-502-N77
ROCK200-502-N77EA 634 EUR
ROCK200-502-N77
Anti-CD8A Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 53-6.7]
Antikörper
Anti-CD8a is useful for immunochemistry, Immunoprecipitation, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.
Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg); Sufficient to run approximately 500 tests
Do not freeze!
Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse