Anti-PTPRC Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: RA3-6B2]

Lieferant: Rockland Immunochemicals
200-502-N84
ROCK200-502-N84EA 699 EUR
ROCK200-502-N84
Anti-PTPRC Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: RA3-6B2]
Antikörper
Anti-CD45R (B220) is useful for Flow Cytometry, Immunoprecipitation, immunochemistry, Immunofluorescence, and Western Blot using mouse spleen cells, or an appropriate cell type (where indicated).

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg); Sufficient to run approximately 500 tests
Do not freeze!

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: RA3-6B2
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR