Anti-CD8A Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 2.43]

Lieferant: Rockland Immunochemicals
200-508-N78
ROCK200-508-N78EA 431 EUR
ROCK200-508-N78
Anti-CD8A Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 2.43]
Antikörper
Anti-CD8a is useful for immunochemistry and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!

Type: Primary
Antigen: CD8A
Clonality: Monoclonal
Clone: 2.43000000000000002
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR