Anti-ITGAM Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: M1/70]

Lieferant: Rockland Immunochemicals
200-508-N79
ROCK200-508-N79EA 873 EUR
ROCK200-508-N79
Anti-ITGAM Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: M1/70]
Antikörper
Anti-CD11b is useful for Immunoprecipitation, immunochemistry, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!

Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: M1/70
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR