Anti-Fcgr3 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 93]

Lieferant: Rockland Immunochemicals
200-508-N80
ROCK200-508-N80EA 944 EUR
ROCK200-508-N80
Anti-Fcgr3 Rat Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 93]
Antikörper
Anti-CD16/32 is useful for Immunoprecipitation and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!

Type: Primary
Antigen: Fcgr3
Clonality: Monoclonal
Clone: 93
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR