Anti-ADGRE1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: BM8.1]

Lieferant: Rockland Immunochemicals
200-526-L55
ROCK200-526-L55EA 856 EUR
ROCK200-526-L55
Anti-ADGRE1 Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: BM8.1]
Antikörper
Anti-F4/80 is useful for Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg)
Do not freeze!

Type: Primary
Antigen: ADGRE1
Clonality: Monoclonal
Clone: BM8.1
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR