Anti-ITGAM Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: M1/70]

Lieferant: Rockland Immunochemicals
200-526-N79
ROCK200-526-N79EA 562 EUR
ROCK200-526-N79
Anti-ITGAM Rat Monoclonal Antibody (APC (Allophycocyanin)) [clone: M1/70]
Antikörper
Anti-CD11b is useful for Immunoprecipitation, immunochemistry, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated).

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze!

Type: Primary
Antigen: ITGAM
Clonality: Monoclonal
Clone: M1/70
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR