Anti-UCHL1 Chicken Polyclonal Antibody

Lieferant: Rockland Immunochemicals
200-901-E52
ROCK200-901-E52EA 861 EUR
ROCK200-901-E52
Anti-UCHL1 Chicken Polyclonal Antibody
Antikörper
Anti-Ubiquitin Hydrolase antibody is suitable for use in ELISA and Western Blotting and IF. Specific conditions for reactivity should be optimized by the end user. Expect a band of approximately 24 kDa in size corresponding to the UCHL1 Proteins in Western blotting.

Recommended Dilutions: ELISA: 1:25000; Immunofluorescence: 1:500; Western Blot: 1:2500

Type: Primary
Antigen: UCHL1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Chicken
Isotype: IgG
Reactivity: Human, Mouse, Rat, Bovine
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR