Anti-IgG F(ab')2 Goat Polyclonal Antibody
Lieferant: Rockland Immunochemicals
202-1104
ROCK202-1104EA 524 EUR
ROCK202-1104
Anti-IgG F(ab')2 Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Cat