Anti-IgG F(c) Donkey Polyclonal Antibody
Lieferant: Rockland Immunochemicals
205-7103
ROCK205-7103EA 526 EUR
ROCK205-7103
Anti-IgG F(c) Donkey Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Donkey
Isotype: IgG
Reactivity: Goat