Anti-IgG (H+L) Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
206-4102
ROCK206-4102EA 617 EUR
ROCK206-4102
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG (Guinea Pig) (Guinea Pig IgG)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestellen