Anti-IL25 Mouse Monoclonal Antibody (Unconjugated) [clone: 11K8B1]
Lieferant: Rockland Immunochemicals
209-301-C58
ROCK209-301-C58EA 841 EUR
ROCK209-301-C58
Anti-IL25 Mouse Monoclonal Antibody (Unconjugated) [clone: 11K8B1]
Antikörper
IL 17E antibody has been tested for use in Flow Cytometry and western blotting. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: ELISA: 1:10000-1:50000; Flow Cytometry: 1µg/ml; Western Blot: 0.5µg/ml;
Type: Primary
Antigen: IL25
Clonality: Monoclonal
Clone: 11K8B1
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human