Anti-PGR Mouse Monoclonal Antibody (Unconjugated) [clone: 1154]

Lieferant: Rockland Immunochemicals
209-301-E16
ROCK209-301-E16EA 980 EUR
ROCK209-301-E16
Anti-PGR Mouse Monoclonal Antibody (Unconjugated) [clone: 1154]
Antikörper
Anti-Progesterone Receptor pS190 (Mouse) antibody is suitable for use in Western Blot and IHC. Expect a band of approximately 90 kDa in size for PR-A isform and approximately 120 kDa in size for the PR-B isoform phosphorylated at Ser190 in Western Blotting.

Recommended Dilutions: Western Blot: 1:1000; Immunochemistry: 1:1000

Type: Primary
Antigen: PGR
Clonality: Monoclonal
Clone: 1154
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR