Anti-HSP90AA1 Mouse Monoclonal Antibody (Unconjugated) [clone: Hyb-K41009]

Lieferant: Rockland Immunochemicals
209-301-F70
ROCK209-301-F70EA 841 EUR
ROCK209-301-F70
Anti-HSP90AA1 Mouse Monoclonal Antibody (Unconjugated) [clone: Hyb-K41009]
Antikörper
Anti-Hsp90 Antibody is suitable for WB, ELISA and IHC. Expect a band approximately 90kDa Proteins corresponding to the molecular mass of Hsp90a on SDS PAGE immunoblots. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200; Western Blot: 1µg/ml

Type: Primary
Antigen: HSP90AA1
Clonality: Monoclonal
Clone: Hyb-K41009
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR