Anti-VEGFA Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
209-401-B99
ROCK209-401-B99EA 821 EUR
ROCK209-401-B99
Anti-VEGFA Rabbit Polyclonal Antibody
Antikörper
This purified antibody has been tested for use in neutralizations, ELISA and western blotting. Reactivity is also expected in radioimmunoassay.

Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000;
Restore with deionized water (or equivalent); 100 µl

Type: Primary
Antigen: VEGFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR