Anti-IL32 Rabbit Polyclonal Antibody (Biotin)
Lieferant: Rockland Immunochemicals
209-406-B49
ROCK209-406-B49EA 791 EUR
ROCK209-406-B49
Anti-IL32 Rabbit Polyclonal Antibody (Biotin)
Antikörper
This purified antibody has been tested for use in ELISA and western blotting. By western blot a band approximately 15 kDa in size corresponding to native human IL-32a Protein is expected in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: 1:20000-1:100000; Western Blot: 1:2000-1:10000; Immunochemistry: 1:1000-1:5000
Restore with deionized water (or equivalent); 100 µl
Type: Primary
Antigen: IL32
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Full length
Host: Rabbit
Isotype: IgG
Reactivity: Human