Anti-Mouse IL-1 beta Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
210-401-319
ROCK210-401-319EA 821 EUR
ROCK210-401-319
Anti-Mouse IL-1 beta Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Mouse IL-1ß
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR