Anti-IgG (H+L) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: Rockland Immunochemicals
213-4202
ROCK213-4202EA 579 EUR
ROCK213-4202
Anti-IgG (H+L) Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)Epitope:
Host: Rabbit
Isotype: none
Reactivity: Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR