Anti-IgG (H+L) Donkey Polyclonal Antibody

Lieferant: Rockland Immunochemicals
213-7102
ROCK213-7102EA 873 EUR
ROCK213-7102
Anti-IgG (H+L) Donkey Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG (Sheep) (sheep IgG)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Donkey
Isotype: none
Reactivity: Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR