Anti-TNFA Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
214-401-306S
ROCK214-401-306SEA 226 EUR
ROCK214-401-306S
Anti-TNFA Rabbit Polyclonal Antibody
Antikörper
This Protein A purified antibody is suitable for ELISA and Western blotting.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 16.9 kDa in size corresponding to swine TNFa by western blotting.

Recommended Dilutions: ELISA: 1:2000 - 1:2500; Western Blot: 1:1000;

Type: Primary
Antigen: TNFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR