Anti-CCL4 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
214-401-B85
ROCK214-401-B85EA 821 EUR
ROCK214-401-B85
Anti-CCL4 Rabbit Polyclonal Antibody
Antikörper
This Protein-A purified antibody is suitable for ELISA and Western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 7.8 kDa in size corresponding to swine CCL4 Protein by western blotting.

Recommended Dilutions: ELISA: >1:5000; Western Blot: 1:500;

Type: Primary
Antigen: CCL4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR