Anti-IgG (H+L) Rabbit Polyclonal Antibody
Lieferant: Rockland Immunochemicals
308-4102
ROCK308-4102EA 464 EUR
ROCK308-4102
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Horse