Anti-Biotin Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))

Lieferant: Rockland Immunochemicals
600-100-098
ROCK600-100-098EA 1080 EUR
ROCK600-100-098
Anti-Biotin Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Antikörper
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions:
Restore with deionized water (or equivalent); 1.0 ml

Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan
Bestellen

TECHNISCHE DATEN

Antigen-Name Biotin
Antikörper-Typ Tag
Klonalität Polyklonal
Konjugation TRITC (Tetramethylrhodamine Isothiocyanate)
Reaktivität Multiple
Wirt Goat
Western blot Yes
ELISA Yes
ImmunoChemie Yes
ImmunFluoreszenz Yes
Immunogen Biotin conjugated to Keyhole Limpet Hemocyanin (KLH)
Format Antibody fraction
Form Lyophilized
Reinigung purified by immunoaffinity chromatography
Konzentration 1,0 mg/ml by UV absorbance at 280 nm
Kreuzadsorption none
Lagerungspuffer 0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
Lagertemperatur Cool
Transporttemperatur Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR