Anti-FITC Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
600-101-096S
ROCK600-101-096SEA 226 EUR
ROCK600-101-096S
Anti-FITC Goat Polyclonal Antibody
Antikörper
Anti-Fluorescein Antibody is suitable for immunomicroscopy, in situ hybridization and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:5000 -1:30000; Western Blot: 1:500 -1:3000; Immunochemistry: 1:250 -1:1500;

Type: Tag
Antigen: FITC
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR