Anti-GFP Goat Polyclonal Antibody
Lieferant: Rockland Immunochemicals
600-101-215
ROCK600-101-215EA 1060 EUR
ROCK600-101-215
Anti-GFP Goat Polyclonal Antibody
Antikörper
Anti-GFP is designed to detect GFP and its variants. Goat Anti-GFP is ideal for western blotting, ELISA, immunochemistry and IP.
Recommended Dilutions: ELISA: 1:20000; Immunofluorescence: 1:500; Western Blot: 1:400 - 1:2000; Immunochemistry: 1:200 - 1:1000; Immunoprecipitation: Yes;
Type: Tag
Antigen: GFP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Goat
Isotype: IgG
Reactivity:
Bestellen