Anti-ING4 Goat Polyclonal Antibody
Lieferant: Rockland Immunochemicals
600-101-284
ROCK600-101-284EA 821 EUR
ROCK600-101-284
Anti-ING4 Goat Polyclonal Antibody
Antikörper
This affinity-purified antibody has been tested for use in ELISA against the immunizing peptide.  Specific conditions for western blotting reactivity should be optimized by the end user. Expect a band at approximately 29 kDa in size corresponding to ING4.
Recommended Dilutions: ELISA: 1:3000 - 1:12000; Western Blot: 1:500 - 1:3000;
Type: Primary
Antigen: ING4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human