Anti-CHAT Goat Polyclonal Antibody
Lieferant: Rockland Immunochemicals
600-101-D16
ROCK600-101-D16EA 976 EUR
ROCK600-101-D16
Anti-CHAT Goat Polyclonal Antibody
Antikörper
Anti-ChAT antibody is suitable for use in ELISA, Western Blotting and IHC. Specific conditions for reactivity should be optimized by the end user. Expect a ~ 68/70 kDa band corresponding to choline acetyltransferase Protein by Western Blotting.
Recommended Dilutions: Western Blot: 1:100; Immunochemistry: 1:100
Type: Primary
Antigen: CHAT
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human, non-Human Primate, Chicken, Guinea Pig, Mouse