Anti-RPE Goat Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Lieferant: Rockland Immunochemicals
600-102-387
ROCK600-102-387EA 1010 EUR
ROCK600-102-387
Anti-RPE Goat Polyclonal Antibody (FITC (Fluorescein isothiocyanate))
Antikörper
Suitable for immunomicroscopy and flow cytometry or FACS analysis as well as other antibody based fluorescent assays requiring lot-to-lot consistency.  .
Recommended Dilutions:
Restore with deionized water (or equivalent); 1.0 ml
Type: Tag
Antigen: RPE
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Bestellen